Anti-TR2IT1 Rabbit Polyclonal Antibody
Catalog # 10273-098
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:TR2IT1
- Antigen symbol:TR2IT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Cross adsorption:No
- Storage buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen affinity purification
- Cat. no.:10273-098
- Supplier No.:25753-1-AP
Specifications
About this item
The TR2IT1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TR2IT1. This antibody recognizes human antigen. The TR2IT1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: TR2IT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human