Anti-TDO2 Rabbit Polyclonal Antibody
Catalog # 10713-764
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Tryptophan 2,3-dioxygenase
- Antigen symbol:TDO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Cross adsorption:No
- Antigen synonyms:TRPO|TO|TDO|TPH2
- Storage buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity purification
- Cat. no.:10713-764
- Supplier No.:15880-1-AP
Specifications
About this item
The TDO2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human TDO2. This antibody recognizes human,mouse,rat antigen. The TDO2 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: TDO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat