4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) solution 1 mg/mL for fluorescence microscopy, used in nuclear staining, Pierce™
CAPI62248
:
- Size:1 ml
- Cat. no.:CAPI62248
- Supplier No.:62248
- Color:Blue
- For Use With (Equipment):Fluorescence Microscope, High Content Instrument
- Cell Permeability:Cell-Impermeant
- Sub-Cellular Localization:Nucleus
- Detection Method:Fluorescent
- Excitation⁄Emission (nm):360⁄460
:
:350.25 g/mol
:28718-90-3
:C₁₆H₁₅N₅·2HCl
:MFCD00012681