Cork Borer Set with Handles, Humboldt
These cork borer sets are used to bore holes in corks and rubber stoppers.
They consist of plated tubing with precision ground cutting edges. The smooth plastic handles are stamped with a number designation. A punch is included with each set.
Frequently Bought Together