Anti-TBK1 Rabbit Polyclonal Antibody
Catalog # 10210-064
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:TANK-binding kinase 1
- Antigen symbol:TBK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:TBK1|Serine/threonine-protein kinase TBK1|NAK|NF-kappa-B-activating kinase|TANK-binding kinase 1|T2K
- Amino acid number:577 to 590
- Sequence:YNEEQIHKFDKQKL
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human NAK (577-590aa YNEEQIHKFDKQKL), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10210-064
- Supplier No.:PA2117
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase TBK1(TBK1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TBK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat