Anti-YB1 Rabbit Polyclonal Antibody
Catalog # 10210-054
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Y box binding Protein 1
- Antigen symbol:YB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CCAAT-binding transcription factor I subunit A|EFI-A|YB1|DBPB|CBF-A|Y-box-binding protein 1|NSEP-1|nuclease-sensitive element-binding protein 1|CSDA2|Y box-binding protein I|DNA-binding protein B|major histocompatibility complex|NSEP1|MDR-NF1|class II|BP-8|MGC110976|enhancer factor I subunit A|MGC117250|CSDB|nuclease sensitive element binding protein 1|YB-1|MGC104858|Y-box transcription factor
- Amino acid number:137 to 152
- Sequence:KYAADRNHYRRYPRRR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human YB1 (137-152aa KYAADRNHYRRYPRRR), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10210-054
- Supplier No.:PA1758
Specifications
About this item
Rabbit IgG polyclonal antibody for Nuclease-sensitive element-binding protein 1(YBX1) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: YB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat