Anti-LTBR Rabbit Polyclonal Antibody
Catalog # 10209-798
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Lymphotoxin beta receptor
- Antigen symbol:LTBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CD18|TNFR2-RP|TNFR-RP|TNFR3|TNF-R-III|TNFCR|LT-BETA-R|TNFRSF3|D12S370
- Amino acid number:420 to 435
- Sequence:ATPSNRGPRNQFITHD
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human LTBR (420-435aa ATPSNRGPRNQFITHD).
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-798
- Supplier No.:PA1968
Specifications
About this item
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 3(LTBR) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: LTBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human