Anti-GJB2 Rabbit Polyclonal Antibody
Catalog # 10209-730
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Gap Junction Protein, Beta 2
- Antigen symbol:GJB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CX26|DFNB1A|DFNB1|NSRD1|HID|DFNA3A|PPK|DFNA3|KID|connexin 26|Gap junction beta-2 protein
- Amino acid number:1 to 16
- Sequence:MDWGTLQTILGGVNKH
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human GJB2 (1-16aa, MDWGTLQTILGGVNKH), different from the related mouse and rat sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-730
- Supplier No.:PA1025
Specifications
About this item
Rabbit IgG polyclonal antibody for Gap junction beta-2 protein(GJB2) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: GJB2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat