Anti-TRAF1 Rabbit Polyclonal Antibody
Catalog # 10209-650
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:TNF Receptor-associated Factor 1
- Antigen symbol:TRAF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:MGC:10353|EBI6
- Amino acid number:220 to 235
- Sequence:HQSQLDRERILSLEQR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human TRAF1 (220-235aa HQSQLDRERILSLEQR), different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-650
- Supplier No.:PA2006
Specifications
About this item
Rabbit IgG polyclonal antibody for TNF receptor-associated factor 1(TRAF1) detection. Tested with WB in Human;Mouse.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TRAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse