Anti-CRTC2 Rabbit Polyclonal Antibody
Catalog # 10209-646
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:CREB-Regulated Transcription Coactivator 2
- Antigen symbol:CRTC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:TORC2|transducer of regulated cAMP response element-binding protein 2|CREB-regulated transcription coactivator 2|TORC-2|transducer of CREB protein 2
- Amino acid number:595 to 611
- Sequence:QDPHTFNHQNLTHCSRH
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TORC2 (595-611aa QDPHTFNHQNLTHCSRH), different from the related rat sequence by three amino acids, and from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-646
- Supplier No.:PA1988
Specifications
About this item
Rabbit IgG polyclonal antibody for CREB-regulated transcription coactivator 2(CRTC2) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CRTC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: