Anti-HDAC8 Rabbit Polyclonal Antibody
Catalog # 10209-566
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HDAC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:18 to 33
- Sequence:YIYSPEYVSMCDSLAK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human HDAC8 (18-33aa YIYSPEYVSMCDSLAK), different from the related mouse and rat sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-566
- Supplier No.:PA1591
Specifications
About this item
Rabbit IgG polyclonal antibody for Histone deacetylase 8(HDAC8) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat