Order Entry
Canada
ContactUsLinkComponent
 
Anti-Caspase-3(P10) Rabbit Polyclonal Antibody
  10209-542
undefined
Anti-Caspase-3(P10) Rabbit Polyclonal Antibody
  10209-542
 :  PA1302-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Caspase 3
  • Antigen symbol:
    CASP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    procaspase3|Yama|CASP-3|CASP3|CPP32|Apopain|CPP32B|SCA-1|CPP-32|apopain
  • Amino acid number:
    220 to 236
  • Sequence:
    CAMLKQYADKLEFMHIL
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Caspase-3 (P10) (220-236aa CAMLKQYADKLEFMHIL).
  • Purification:
    purified by immunogen affinity purification
  • Cat. no.:
    10209-542
  • Supplier No.:
    PA1302-1

 

 

Rabbit IgG polyclonal antibody for Caspase-3(CASP3) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Caspase-3(P10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human