Anti-CXCR2 Rabbit Polyclonal Antibody
Catalog # 10209-522
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C Motif) Receptor 2
- Antigen symbol:CXCR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:IL8RB|CD182|IL8R2|CDw128b|IL8RA|mIL-8RH|Il8rb|CMKAR2|IL-8Rh|IL-8rb
- Amino acid number:196 to 212
- Sequence:CYEDMGNNTANWRMLLR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human CXCR2 (196-212aa CYEDMGNNTANWRMLLR), different from the related mouse sequence by six amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-522
- Supplier No.:PA1029
Specifications
About this item
Rabbit IgG polyclonal antibody for C-X-C chemokine receptor type 2(CXCR2) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CXCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: