Anti-MATN2 Rabbit Polyclonal Antibody
10209-388
- Antibody type:Primary
- Antigen name:Matrilin 2
- Antigen symbol:MATN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:466 to 486
- Sequence:NTKLDDILEKLKINEYGQIHR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Matrilin 3 (466-486aa NTKLDDILEKLKINEYGQIHR).
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-388
- Supplier No.:PA1927
Rabbit IgG polyclonal antibody for Matrilin-3(MATN3) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: MATN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human