Anti-COL2 Rabbit Polyclonal Antibody
Catalog # 10209-300
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Collagen II
- Antigen symbol:COL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:chondrocalcin|cartilage collagen|collagen II|COL2A1|alpha-1 type II collagen|Type II Collagen|Collagen 2|collagen alpha-1(II) chain|alpha-1 polypeptide
- Amino acid number:1222 to 1241
- Sequence:AFAGLGQREKGPDPMQYMRA
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Collagen II1222-1241aa AFAGLGQREKGPDPMQYMRA), different from the related rat sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-300
- Supplier No.:PA2141-1
Specifications
About this item
Rabbit IgG polyclonal antibody for Collagen alpha-1(II) chain(COL2A1) detection. Tested with WB, IHC-F in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: COL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: