Anti-CTNNB1 Rabbit Polyclonal Antibody
Catalog # 10209-234
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Catenin beta
- Antigen symbol:CTNNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Beta-catenin|ORF Names: OK/SW-cl.35|Catenin beta-1|PRO2286|Catenin beta|CTNNB|CTNNB1
- Amino acid number:1 to 19
- Sequence:MATQADLMELDMAMEPDRK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human beta Catenin (1-19aa MATQADLMELDMAMEPDRK), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-234
- Supplier No.:PA1212-1
Specifications
About this item
Rabbit IgG polyclonal antibody for Catenin beta-1(CTNNB1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CTNNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: