Anti-Glutaredoxin 2 Rabbit Polyclonal Antibody
Catalog # 10209-178
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Glutaredoxin 2
- Antigen symbol:GLRX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:103 to 119
- Sequence:EYGNQFQDALYKMTGER
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human Glutaredoxin 2 (103-119aa EYGNQFQDALYKMTGER), different from the related rat sequence by two amino acids, and from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10209-178
- Supplier No.:PA1885
Specifications
About this item
Rabbit IgG polyclonal antibody for Glutaredoxin-2, mitochondrial(GLRX2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Glutaredoxin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat