Order Entry
Canada
ContactUsLinkComponent
Anti-Estrogen Inducible Protein pS2 Rabbit Polyclonal Antibody
Anti-Estrogen Inducible Protein pS2 Rabbit Polyclonal Antibody
Catalog # 10209-090
undefined
Anti-Estrogen Inducible Protein pS2 Rabbit Polyclonal Antibody
Catalog # 10209-090
Supplier Number:  PA1690

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Estrogen Inducible Protein pS2
  • Antigen symbol:
    TFF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    59 to 72
  • Sequence:
    CCFDDSVRGFPWCF
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of mouse Estrogen Inducible Protein pS2 (59-72aa CCFDDSVRGFPWCF), identical to the related rat sequence.
  • Purification:
    purified by immunogen affinity purification
  • Cat. no.:
    10209-090
  • Supplier No.:
    PA1690

Specifications

About this item

Rabbit IgG polyclonal antibody for Trefoil factor 1(TFF1) detection. Tested with WB, IHC-P in Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Estrogen Inducible Protein pS2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat