Anti-IgG Antibody (HRP (Horseradish Peroxidase))
10208-926
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- Cross adsorption:No
- Cat. no.:10208-926
- Supplier No.:BA1058-1
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity: