Anti-IgG Antibody (Biotin)
Catalog # 10208-890
Supplier: Boster Biological Technology
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Biotin
- Cross adsorption:No
- Cat. no.:10208-890
- Supplier No.:BA1005-0.5
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: