Anti-IgM Antibody (Biotin)
Catalog # 10208-888
Supplier: Boster Biological Technology
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin M
- Antigen symbol:IgM
- Conjugation:Biotin
- Cross adsorption:No
- Cat. no.:10208-888
- Supplier No.:BA1004-0.25
Specifications
About this item
Type: Secondary
Antigen: IgM
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: