Anti-LPP Rabbit Polyclonal Antibody
Catalog # 10208-852
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:LIM Domain Containing Preferred Translocation Partner In Lipoma
- Antigen symbol:LPP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:291 to 307
- Sequence:PNQGRYYEGYYAAGPGY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human LPP (291-307aa PNQGRYYEGYYAAGPGY).
- Purification:purified by immunogen affinity purification
- Cat. no.:10208-852
- Supplier No.:PA2040
Specifications
About this item
Rabbit IgG polyclonal antibody for Lipoma-preferred partner(LPP) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: LPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human