Anti-VGF Rabbit Polyclonal Antibody
Catalog # 10208-840
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:VGF nerve growth Factor inducible
- Antigen symbol:VGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Neurosecretory protein VGF|VGF|NERP-1
- Amino acid number:582 to 600
- Sequence:QARRAQEEAEAEERRLQEQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human VGF (582-600aa QARRAQEEAEAEERRLQEQ), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10208-840
- Supplier No.:PA1984
Specifications
About this item
Rabbit IgG polyclonal antibody for Neurosecretory protein VGF(VGF) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: VGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat