Anti-REA Rabbit Polyclonal Antibody
10208-836
- Antibody type:Primary
- Antigen symbol:REA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:111 to 128
- Sequence:RPNAQELPSMYQRLGLDY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human REA (111-128aa RPNAQELPSMYQRLGLDY), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10208-836
- Supplier No.:PA1951
Rabbit IgG polyclonal antibody for Prohibitin-2(PHB2) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: REA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat