Anti-HSP70 Mouse Monoclonal Antibody [clone: SJ-70]
Catalog # 10207-946
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein 70
- Antigen symbol:HSP70
- Clonality:Monoclonal
- Clone:SJ-70
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Bovine,Rat
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- Storage temperature:–20 °C
- Immunogen:HSP70 isolated from bovine brain.
- Cat. no.:10207-946
- Supplier No.:MA1050
Specifications
About this item
Mouse IgG monoclonal antibody for HSP70, heat shock 70kDa protein 1A; heat shock 70kDa protein 1B (HSPA1A; HSPA1B) detection. Tested with WB, IHC-P, IHC-F in Human. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: SJ-70
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Rabbit, Bovine