Order Entry
Canada
ContactUsLinkComponent
 
Anti-HSP25 Mouse Monoclonal Antibody [clone: SJ-25]
  10207-942
Anti-HSP25 Mouse Monoclonal Antibody [clone: SJ-25]
  10207-942
 :  MA1048

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat Shock Protein 25
  • Antigen symbol:
    HSP25
  • Clonality:
    Monoclonal
  • Clone:
    SJ-25
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rabbit,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    Stress-responsive protein 27|Heat shock 27 kDa protein|SRP27|HSP27|HspB1|HSP25|Estrogen-regulated 24 kDa protein|HSPB1|HSP 27|Heat shock protein beta-1|28 kDa heat shock protein
  • Storage temperature:
    –20 °C
  • Immunogen:
    Partially purified inhibitor of actin polymerization (IAP) protein from turkey gizzard smooth muscle.
  • Cat. no.:
    10207-942
  • Supplier No.:
    MA1048

 

 

Mouse IgG monoclonal antibody for HSP25, heat shock 27kDa protein 1 (HSPB1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human. No cross reactivity with other proteins.

Reconstitute with 1.2% sodium acetate or neutral PBS.

If 1ml of PBS is used, the antibody concentration will be 100ug/ml.

Type: Primary
Antigen: HSP25
Clonality: Monoclonal
Clone: SJ-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Rabbit