Anti-DES Mouse Monoclonal Antibody [clone: DES-82]
Catalog # 10207-938
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Desmin
- Antigen symbol:DES
- Clonality:Monoclonal
- Clone:DES-82
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CSM1|SCPNK|MFM1|CMD1I|intermediate filament protein|CSM2
- Storage temperature:–20 °C
- Immunogen:Desmin from pig stomach.
- Cat. no.:10207-938
- Supplier No.:MA1036
Specifications
About this item
Mouse IgG monoclonal antibody for Desmin , desmin (DES) detection. Tested with WB, IHC-P in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: DES-82
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Rabbit