Order Entry
Canada
ContactUsLinkComponent
Anti-Kv1.1 potassium channel Rabbit Polyclonal Antibody
Anti-Kv1.1 potassium channel Rabbit Polyclonal Antibody
Catalog # 10207-928
undefined
Anti-Kv1.1 potassium channel Rabbit Polyclonal Antibody
Catalog # 10207-928
Supplier Number:  PA2296

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Kv1.1 potassium channel
  • Antigen symbol:
    KV1.1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    465 to 481
  • Sequence:
    IAHYRQVNIRTANCTTA
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Kv1.1 potassium channel (465-481aa IAHYRQVNIRTANCTTA), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids .
  • Purification:
    purified by immunogen affinity purification
  • Cat. no.:
    10207-928
  • Supplier No.:
    PA2296

Specifications

About this item

Rabbit IgG polyclonal antibody for Potassium voltage-gated channel subfamily A member 1(KCNA1) detection. Tested with WB in Human;Mouse.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Kv1.1 potassium channel
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse