Order Entry
Canada
ContactUsLinkComponent
Anti-Avidin Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-Avidin Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # 10207-570
undefined
Anti-Avidin Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # 10207-570
Supplier Number:  BA1093-1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Avidin
  • Antigen symbol:
    Avidin
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • Cross adsorption:
    No
  • Antigen synonyms:
    AVD
  • Cat. no.:
    10207-570
  • Supplier No.:
    BA1093-1

Specifications

About this item

Type: Primary
Antigen: Avidin
Clonality:
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: 0
Isotype:
Reactivity: 0