Anti-MAP2K4 Rabbit Polyclonal Antibody
Catalog # 10207-434
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Dual specificity mitogen-activated protein kinase kinase 4
- Antigen symbol:MAP2K4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:MKK4,|Dual specificity mitogen-activated protein kinase kinase 4|SAPKK-1|PRKMK4|MEK 4|SAPKK1|MAPKK 4|MAP2K4|SAPK/ERK kinase 1|c-Jun N-terminal kinase kinase 1|SERK1|SKK1|MEK4|JNKK1|JNK-activating kinase 1|JNKK|SEK1|Stress-activated protein kinase kinase 1|SAPK kinase 1|MAP kinase kinase 4|MAPK/ERK kinase 4
- Amino acid number:360 to 378
- Sequence:ELLKHPFILMYEERAVEVA
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MEK4 (360-378aa ELLKHPFILMYEERAVEVA), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-434
- Supplier No.:PA2275
Specifications
About this item
Rabbit IgG polyclonal antibody for Dual specificity mitogen-activated protein kinase kinase 4(MAP2K4) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat