Anti-KLF4 Rabbit Polyclonal Antibody
10207-418
- Antibody type:Primary
- Antigen name:Kruppel-like Factor 4
- Antigen symbol:KLF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:93 to 108
- Sequence:RRETEEFNDLLDLDFI
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human KLF4 (93-108aa RRETEEFNDLLDLDFI), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-418
- Supplier No.:PA2211
Rabbit IgG polyclonal antibody for Krueppel-like factor 4(KLF4) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: KLF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat