Anti-IRF5 Rabbit Polyclonal Antibody
10207-414
- Antibody type:Primary
- Antigen name:IRF5
- Antigen symbol:IRF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:456 to 472
- Sequence:EQFKELHHIWQSQQRLQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human IRF5 (456-472aa EQFKELHHIWQSQQRLQ), identical to the related rat sequence, and different from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-414
- Supplier No.:PA2208
Rabbit IgG polyclonal antibody for Interferon regulatory factor 5(IRF5) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: IRF5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat