Anti-PDK2 Rabbit Polyclonal Antibody
10207-334
- Antibody type:Primary
- Antigen name:Pyruvate Dehydrogenase Kinase 2
- Antigen symbol:PDK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Pyruvate dehydrogenase kinase isoform 2|pyruvate dehydrogenase kinase|PDK2_HUMAN.|mitochondrial|PDHK2|Pyruvate dehydrogenase lipoamide kinase isozyme 2|isozyme 2
- Amino acid number:379 to 392
- Sequence:WRHYQTIQEAGDWC
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human PDK2 (379-392aa WRHYQTIQEAGDWC), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-334
- Supplier No.:PA1994
Rabbit IgG polyclonal antibody for[Pyruvate dehydrogenase[lipoamide]] kinase isozyme 2, mitochondrial(PDK2) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: PDK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat