Anti-TFPI Rabbit Polyclonal Antibody
10207-284
- Antibody type:Primary
- Antigen name:Tissue Factor Pathway Inhibitor
- Antigen symbol:TFPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:TFI|EPI|lipoprotein-associated coagulation inhibitor|LACI|tissue factor pathway inhibitor|anti-convertin|TFPI1|extrinsic pathway inhibitor
- Amino acid number:261 to 280
- Sequence:QECLRACKKGFIQRISKGGL
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human TFPI (261-280aa QECLRACKKGFIQRISKGGL).
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-284
- Supplier No.:PA1798
Rabbit IgG polyclonal antibody for Tissue factor pathway inhibitor(TFPI) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TFPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human