Order Entry
Canada
ContactUsLinkComponent
 
Anti-HEXA Rabbit Polyclonal Antibody
Catalog # 10207-278
undefined
Anti-HEXA Rabbit Polyclonal Antibody
Catalog # 10207-278
Supplier Number:  PA1787-2

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HEXA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    513 to 529
  • Sequence:
    QAQPLNVGFCEQEFEQT
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human HEXA (513-529aa QAQPLNVGFCEQEFEQT), different from the related mouse sequence by three amino acids, and from the related rat sequences by four amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Cat. no.:
    10207-278
  • Supplier No.:
    PA1787-2

Specifications

About this item

Rabbit IgG polyclonal antibody for Beta-hexosaminidase subunit alpha(HEXA) detection. Tested with WB in Human.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: HEXA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human