Anti-Ku80 Rabbit Polyclonal Antibody
Catalog # 10207-210
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:5'-deoxyribose-5-phosphate lyase Ku80
- Antigen symbol:Ku80
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:complem???????|Ku autoantigen|CTC85|TLAA|Ku86 autoantigen related protein 1,TL86 autoantigen related protein 1|DNA repair protein XRCC5|ATP-dependent DNA helicase 2 subunit 2|86 kDa subunit of Ku antigen|80kD)|X-ray repair cross-complementing protein 5|X-ray repair|CTCBF|ATP-dependent DNA helicase II 80 kDa subunit|X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining|CTC box-binding factor 85 kDa subunit|80kDa
- Amino acid number:648 to 662
- Sequence:KFSEEQRFNNFLKAL
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Ku80 (648-662aa KFSEEQRFNNFLKAL).
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-210
- Supplier No.:PA1641
Specifications
About this item
Rabbit IgG polyclonal antibody for X-ray repair cross-complementing protein 5(XRCC5) detection. Tested with WB, IHC-P, ICC in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Ku80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human