Anti-CCR4 Rabbit Polyclonal Antibody
10207-172
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 4
- Antigen symbol:CCR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ChemR13|CC-CKR-4|CKR4|HGCN:14099|K5-5|CMKBR4|CD194
- Amino acid number:346 to 360
- Sequence:SYTQSTVDHDFRDAL
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of mouse CCR4 (346-360aa, SYTQSTVDHDFRDAL), identical to the related rat sequence.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-172
- Supplier No.:PA1517
Rabbit IgG polyclonal antibody for C-C chemokine receptor type 4(CCR4) detection. Tested with WB in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat