Anti-IL6 Rabbit Polyclonal Antibody
Catalog # 10207-138
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 6
- Antigen symbol:IL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:HSF|HGF|IFNB2|interferon beta 2|BSF2|IL-6
- Amino acid number:195 to 211
- Sequence:KALEEFLKVTMRSTRQT
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat IL6 (195-211aa KALEEFLKVTMRSTRQT), different from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-138
- Supplier No.:PA1352
Specifications
About this item
Rabbit IgG polyclonal antibody for Interleukin-6(IL6) detection. Tested with WB in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat