Anti-CHK2 Rabbit Polyclonal Antibody
10207-104
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 2
- Antigen symbol:CHK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:PP1425|LFS2|RAD53|CDS1|OTTHUMP00000028871|bA444G7|HuCds1|CHK2|Cds1
- Amino acid number:427 to 441
- Sequence:FSEHRTQVSLKDQIT
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Chk2 (427-441aa FSEHRTQVSLKDQIT), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-104
- Supplier No.:PA1202
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase Chk2(CHEK2) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CHK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: