Anti-SKP2 Rabbit Polyclonal Antibody
Catalog # 10207-090
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:S-phase Kinase-associated Protein 2
- Antigen symbol:SKP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SKP2|FLB1|MGC1366|FBL1|FBXL1
- Amino acid number:2 to 20
- Sequence:HRKHLQEIPDLSSNVATSF
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human SKP2 (2-20aa HRKHLQEIPDLSSNVATSF), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-090
- Supplier No.:PA1102
Specifications
About this item
Rabbit IgG polyclonal antibody for S-phase kinase-associated protein 2(SKP2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: SKP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat