Anti-RUNX3 Rabbit Polyclonal Antibody
Catalog # 10207-008
Supplier: Boster Biological Technology
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 3
- Antigen symbol:RUNX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:AML2|Core-binding factor subunit alpha-3|RUNX3|SL3-3 enhancer factor 1 alpha C subunit|PEBP2A3|Runt-related transcription factor 3|Oncogene AML-2|CBFA3|Polyomavirus enhancer-binding protein 2 alpha C subunit|Acute myeloid leukemia 2 protein|SL3/
- Amino acid number:186 to 205
- Sequence:QKIEDQTKAFPDRFGDLRMR
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of mouse RUNX3 (186-205aa QKIEDQTKAFPDRFGDLRMR), identical to the related rat sequence.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-008
- Supplier No.:PA2112
Specifications
About this item
Rabbit IgG polyclonal antibody for Runt-related transcription factor 3(RUNX3) detection. Tested with WB in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat