Anti-ABCB4 Rabbit Polyclonal Antibody
10207-002
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, Subfamily B, Member 4
- Antigen symbol:ABCB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:P-glycoprotein 3|PGY3|ABC21|MDR2/3|MDR3|MDR2|GBD1|ICP3|Multidrug resistance protein 3|PFIC-3
- Amino acid number:1 to 20
- Sequence:MDLEAAKNGTAWRPTSAEGD
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human ABCB4 (1-20aa MDLEAAKNGTAWRPTSAEGD).
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-002
- Supplier No.:PA2090
Rabbit IgG polyclonal antibody for Multidrug resistance protein 3(ABCB4) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: ABCB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human