Anti-DISC1 Rabbit Polyclonal Antibody
10206-976
- Antibody type:Primary
- Antigen name:Disrupted in schizophrenia 1
- Antigen symbol:DISC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SCZD9|KIAA0457|C1orf136
- Amino acid number:461 to 477
- Sequence:EKQQLQKEIEALQARMF
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human DISC1 (461-477aa EKQQLQKEIEALQARMF), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-976
- Supplier No.:PA2023-1
Rabbit IgG polyclonal antibody for Disrupted in schizophrenia 1 protein(DISC1) detection. Tested with WB, IHC-P in Human;Mouse.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: DISC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse