Anti-TRAM2 Rabbit Polyclonal Antibody
Catalog # 10206-970
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Translocation Associated Membrane Protein 2
- Antigen symbol:TRAM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:309 to 325
- Sequence:RFIHSQLRHWREYWKEQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse TRAM2 (309-325aa RFIHSQLRHWREYWKEQ), identical to the related rat sequence, and different from the related human sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-970
- Supplier No.:PA2011
Specifications
About this item
Rabbit IgG polyclonal antibody for Translocating chain-associated membrane protein 2(TRAM2) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TRAM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat