Anti-DUSP3 Rabbit Polyclonal Antibody
100 µg
Catalog # 10206-912
Supplier: Boster Biological Technology
Supplier #: PA1762
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit IgG polyclonal antibody for Dual specificity protein phosphatase 3(DUSP3) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: DUSP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
PA1762
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by immunogen affinity purification
- Antibody type:Primary
- Epitope:near the C-terminus
- Form:Lyophilized
- Amino acid number:155 to 171
- Isotype:IgG
- Storage temperature:–20 °C
- Host:Rabbit
- Sequence:RQNREIGPNDGFLAQLC
- Cross adsorption:No
- Antigen name:Dual Specificity Phosphatase 3
- Cat. No.:10206-912
- Reactivity:Human|Rat|Mouse
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human DUSP3 (155-171aa RQNREIGPNDGFLAQLC), identical to the related rat and mouse sequences.
- Conjugation:Unconjugated
- Antigen symbol:DUSP3



