Anti-STAT4 Rabbit Polyclonal Antibody
Catalog # 10206-904
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 4
- Antigen symbol:STAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SOCS6|STATI4|SLEB11|STAI4|SOCS4|HSPC060|Signal Transducer and Activator of Transcription|CIS4|SSI4
- Amino acid number:1 to 17
- Sequence:MSQWNQVQQLEIKFLEQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human STAT4 (1-17aa MSQWNQVQQLEIKFLEQ), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-904
- Supplier No.:PA1692
Specifications
About this item
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 4(STAT4) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: STAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat