Anti-TIM1 Rabbit Polyclonal Antibody
Catalog # 10206-880
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis A Virus Cellular Receptor 1
- Antigen symbol:TIM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Tim-1
- Amino acid number:332 to 348
- Sequence:IKALQNAVEKEVQAEDN
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TIM 1 (332-348aa IKALQNAVEKEVQAEDN).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-880
- Supplier No.:PA1624
Specifications
About this item
Rabbit IgG polyclonal antibody for Hepatitis A virus cellular receptor 1(HAVCR1) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human