Anti-FGF22 Rabbit Polyclonal Antibody
10206-830
- Antibody type:Primary
- Antigen name:Fibroblast Growth Factor 22
- Antigen symbol:FGF22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:93 to 109
- Sequence:FYVAMNRRGRLYGSRLY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human FGF22 (93-109aa FYVAMNRRGRLYGSRLY), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-830
- Supplier No.:PA1455
Rabbit IgG polyclonal antibody for Fibroblast growth factor 22(FGF22) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: FGF22
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat