Anti-TXNRD2 Rabbit Polyclonal Antibody
Catalog # 10206-728
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Thioredoxin Reductase 2
- Antigen symbol:TXNRD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SELZ|TR3|TRXR2|TR-BETA|TR
- Amino acid number:418 to 434
- Sequence:RHGQEHVEVYHAHYKPL
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TXNRD2 (418-434aa RHGQEHVEVYHAHYKPL), different from the related rat sequence by three amino acids, and from the related mouse sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-728
- Supplier No.:PA1903
Specifications
About this item
Rabbit IgG polyclonal antibody for Thioredoxin reductase 2, mitochondrial(TXNRD2) detection. Tested with WB, IHC-P in Human;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: TXNRD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat