Anti-STAT5B Rabbit Polyclonal Antibody
10206-720
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 5B
- Antigen symbol:STAT5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:STAT5|Signal transducer and activator of transcription 5B|Transcription factor STAT5B|Stat5b.|STA5B_HUMAN
- Amino acid number:767 to 787
- Sequence:RRVEELLGRPMDSQWIPHAQS
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human STAT5b (767-787aa RRVEELLGRPMDSQWIPHAQS), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-720
- Supplier No.:PA1841
Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 5B(STAT5B) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: STAT5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat